accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHClONH

CAS RN 4638-48-6

IUPAC Name: 5-chloro-2-hydroxy-N-phenylbenzamide
Synonyms: 5-chlorosalicylanilide
Molecular Weight (g/mol): 247.68
Molecular Formula: C13H10ClNO2
InChi Key: KGYNGVVNFRUOOZ-UHFFFAOYSA-N
SMILES: OC1=CC=C(Cl)C=C1C(=O)NC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results