Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chlorosalicylanilide 98.0+%, TCI America™
SDP

Catalog No. C028625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C028625G 25 g
C0286250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C028625G Supplier TCI America Supplier No. C028625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4638-48-6
Molecular Formula C13H10ClNO2
Molecular Weight (g/mol) 247.678
MDL Number MFCD00002320
InChI Key KGYNGVVNFRUOOZ-UHFFFAOYSA-N
PubChem CID 14869
IUPAC Name 5-chloro-2-hydroxy-N-phenylbenzamide
SMILES C1=CC=C(C=C1)NC(=O)C2=C(C=CC(=C2)Cl)O

Specifications

Melting Point 206°C
Color White
Quantity 25 g
Formula Weight 247.68
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chlorosalicylanilide
RTECSNumber CV2450475
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.