accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)(S)OH(R)(R)H3C(R)(S)OHOO

CAS RN 465-21-4

IUPAC Name: 5-[(1R,3aS,5aR,7S,9aS,11aR)-3a,7-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]-2H-pyran-2-one
Synonyms: bufalin
Molecular Weight (g/mol): 386.53
Molecular Formula: C24H34O4
InChi Key: QEEBRPGZBVVINN-ZXRSHIDQSA-N
SMILES: C[C@]12CCC3C(CC[C@@H]4C[C@@H](O)CC[C@]34C)[C@@]1(O)CC[C@@H]2C1=COC(=O)C=C1
Form 
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results