Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Bufalin

Catalog No. 20390010MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-390-010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-390-010MG Supplier MilliporeSigma™ Supplier No. 20390010MG
Add to Cart
Add to Cart

A cardiotonic steroid isolated from toads of Bufo species that potently inhibits ouabain-sensitive Na+,K+-ATPase activity (IC50 = 1.4 nM).

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White to Off-white
Description 98% by HPLC
Target Proteins Ouabain-sensitive Na+,K+-ATPase activity
CAS 465-21-4
Molecular Formula C24H34O4
Formula Weight 386.5g/mol
Solubility DMSO or warm EtOH
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.