accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONHOOOHO

CAS RN 4652-65-7

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}-5-methoxy-5-oxopentanoic acid
Synonyms: 2-{[(benzyloxy)carbonyl]amino}-5-methoxy-5-oxopentanoic acid
Molecular Weight (g/mol): 295.29
Molecular Formula: C14H17NO6
InChi Key: JSNVOAWDAFVIKY-UHFFFAOYNA-N
SMILES: COC(=O)CCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results