Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl N-Carbobenzoxy-L-glutamate 98.0+%, TCI America™
SDP

Catalog No. M22695G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M22695G Supplier TCI America Supplier No. M22695G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 4652-65-7
Melting Point 70°C
Color White
Molecular Formula C14H17NO6
Quantity 5 g
MDL Number MFCD00191090
Synonym 5-Methyl N-Cbz-L-glutamate, N-Carbobenzoxy-L-glutamic Acid 5-Methyl Ester, N-Cbz-L-glutamic Acid 5-Methyl Ester, Z-Glu(OMe)-OH
InChI Key JSNVOAWDAFVIKY-UHFFFAOYNA-N
SMILES COC(=O)CCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
IUPAC Name 2-{[(benzyloxy)carbonyl]amino}-5-methoxy-5-oxopentanoic acid
Molecular Weight (g/mol) 295.29
PubChem CID 6350584
Formula Weight 295.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methyl N-Carbobenzoxy-L-glutamate
Show More Show Less
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.