CAS RN 4655-62-3
IUPAC Name:
1,4-dihydroxy-5-nitronaphthalene-2,3-dicarbonitrile
Synonyms:
1,4-dihydroxy-5-nitronaphthalene-2,3-dicarbonitrile
Molecular Weight (g/mol):
255.19
Molecular Formula:
C12H5N3O4
InChi Key:
FFSZGCAJTTUCCU-UHFFFAOYSA-N
SMILES:
OC1=C(C#N)C(C#N)=C(O)C2=C(C=CC=C12)[N+]([O-])=O