Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dicyano-1,4-dihydroxy-5-nitronaphthalene 98.0+%, TCI America™
SDP

Catalog No. D24345G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D24345G Supplier TCI America Supplier No. D24345G
Add to Cart
Add to Cart

Specifications

CAS 4655-62-3
Molecular Formula C12H5N3O4
Quantity 5 g
MDL Number MFCD00671523
UN Number 3439
Synonym 2,3-Dicyano-5-nitro-1,4-naphthalenediol
InChI Key FFSZGCAJTTUCCU-UHFFFAOYSA-N
SMILES C1=CC2=C(C(=C(C(=C2C(=C1)[N+](=O)[O-])O)C#N)C#N)O
IUPAC Name 1,4-dihydroxy-5-nitronaphthalene-2,3-dicarbonitrile
Molecular Weight (g/mol) 255.189
PubChem CID 12373187
Formula Weight 255.19
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dicyano-1,4-dihydroxy-5-nitronaphthalene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.