accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClOCl

CAS RN 4659-47-6

IUPAC Name: 2,3,6-trichlorobenzaldehyde
Synonyms: 2,3,6-trichlorobenzaldehyde
Molecular Weight (g/mol): 209.45
Molecular Formula: C7H3Cl3O
InChi Key: AURSMWWOMOVHBM-UHFFFAOYSA-N
SMILES: ClC1=CC=C(Cl)C(C=O)=C1Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results