Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,6-Trichlorobenzaldehyde 97.0+%, TCI America™
SDP

Catalog No. T11835G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T11835G Supplier TCI America Supplier No. T11835G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4659-47-6
Molecular Formula C7H3Cl3O
Molecular Weight (g/mol) 209.45
MDL Number MFCD00010216
InChI Key AURSMWWOMOVHBM-UHFFFAOYSA-N
Synonym benzaldehyde, 2,3,6-trichloro, unii-q065lqh643, 2,3,6-trichloro benzaldehyde, pubchem17003, acmc-1apnu, 2,3,6-trichlorbenzaldehyd, 4-07-00-00577 beilstein handbook reference, ksc497o2d, aurora 4807, benzaldehyde,2,3,6-trichloro
PubChem CID 20781
IUPAC Name 2,3,6-trichlorobenzaldehyde
SMILES C1=CC(=C(C(=C1Cl)C=O)Cl)Cl

Specifications

Melting Point 88°C
Quantity 5 g
UN Number 1759
Formula Weight 209.45
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,6-Trichlorobenzaldehyde
RTECSNumber CU8402000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.