accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4663-94-9

IUPAC Name: pyridine-2,3-dicarboxamide
Synonyms: pyridine-2,3-dicarboxamide
Molecular Weight (g/mol): 165.15
Molecular Formula: C7H7N3O2
InChi Key: CJVCXRMYJNMDTP-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(N=CC=C1)C(N)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results