Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine-2,3-dicarboxamide 98.0+%, TCI America™
SDP

Catalog No. P12155G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12155G 5 g
P121525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12155G Supplier TCI America Supplier No. P12155G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4663-94-9
Molecular Formula C7H7N3O2
Molecular Weight (g/mol) 165.152
MDL Number MFCD00191488
InChI Key CJVCXRMYJNMDTP-UHFFFAOYSA-N
PubChem CID 578921
IUPAC Name pyridine-2,3-dicarboxamide
SMILES C1=CC(=C(N=C1)C(=O)N)C(=O)N

Specifications

Color White
Quantity 5 g
Formula Weight 165.15
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Pyridine-2,3-dicarboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.