CAS RN 4667-99-6
CAS RN 4667-99-6
IUPAC Name:
chlorotriethoxysilane
Synonyms:
chloro-triethoxy silane
Molecular Weight (g/mol):
198.72
Molecular Formula:
C6H15ClO3Si
InChi Key:
JEZFASCUIZYYEV-UHFFFAOYSA-N
SMILES:
CCO[Si](Cl)(OCC)OCC
1
–
2
of
2
results
1
Chlorotriethoxysilane 95.0+%, TCI America™
CAS: 4667-99-6 Molecular Formula: C6H15ClO3Si Molecular Weight (g/mol): 198.718 MDL Number: MFCD00015124 InChI Key: JEZFASCUIZYYEV-UHFFFAOYSA-N PubChem CID: 78398 IUPAC Name: chloro(triethoxy)silane SMILES: CCO[Si](OCC)(OCC)Cl
2
Sigma Aldrich Chlorotriethoxysilane
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.