Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorotriethoxysilane 95.0+%, TCI America™
SDP

Catalog No. C33545G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C33545G 5 g
C335425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C33545G Supplier TCI America Supplier No. C33545G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4667-99-6
Molecular Formula C6H15ClO3Si
Molecular Weight (g/mol) 198.718
MDL Number MFCD00015124
InChI Key JEZFASCUIZYYEV-UHFFFAOYSA-N
PubChem CID 78398
IUPAC Name chloro(triethoxy)silane
SMILES CCO[Si](OCC)(OCC)Cl

Specifications

Color Colorless
Boiling Point 157°C
Quantity 5 g
UN Number 2986
Formula Weight 198.72
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Chlorotriethoxysilane
EINECSNumber (2)-2043
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.