accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHONHOO

CAS RN 4668-42-2

IUPAC Name: 3-{[(benzyloxy)carbonyl]amino}-4-methoxy-4-oxobutanoic acid
Synonyms: 3-{[(benzyloxy)carbonyl]amino}-4-methoxy-4-oxobutanoic acid
Molecular Weight (g/mol): 281.26
Molecular Formula: C13H15NO6
InChi Key: MFFFBNAPQRDRQW-UHFFFAOYNA-N
SMILES: COC(=O)C(CC(O)=O)NC(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results