Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl N-Carbobenzoxy-L-aspartate 98.0+%, TCI America™
SDP

Catalog No. M27075G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27075G 5 g
M270725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27075G Supplier TCI America Supplier No. M27075G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4668-42-2
Molecular Formula C13H15NO6
Molecular Weight (g/mol) 281.264
MDL Number MFCD00083277
InChI Key MFFFBNAPQRDRQW-JTQLQIEISA-N
Synonym z-asp-ome, cbz-asp-ome, s-3-benzyloxy carbonyl amino-4-methoxy-4-oxobutanoic acid, 3s-3-benzyloxy carbonyl amino-4-methoxy-4-oxobutanoic acid, 3s-4-methoxy-4-oxo-3-phenylmethoxycarbonylamino butanoic acid, n-cbz-l-aspartic acid 1-methyl ester, n-cbz-l-asp-ome, z-l-asp-ome, pubchem18988
PubChem CID 7021783
IUPAC Name (3S)-4-methoxy-4-oxo-3-(phenylmethoxycarbonylamino)butanoic acid
SMILES COC(=O)C(CC(=O)O)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 91°C
Color White
Quantity 5 g
Formula Weight 281.26
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl N-Carbobenzoxy-L-aspartate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.