accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO

CAS RN 4669-04-9

IUPAC Name: 1-phenylpentadecan-1-one
Synonyms: 1-phenylpentadecan-1-one
Molecular Weight (g/mol): 302.5
Molecular Formula: C21H34O
InChi Key: SDNCECLJQYEDRK-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCC(=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results