Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentadecanophenone 98.0+%, TCI America™
SDP

Catalog No. P19035G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P19035G Supplier TCI America Supplier No. P19035G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4669-04-9
Molecular Formula C21H34O
Molecular Weight (g/mol) 302.50
InChI Key SDNCECLJQYEDRK-UHFFFAOYSA-N
Synonym Phenyl Tetradecyl Ketone
PubChem CID 259220
IUPAC Name 1-phenylpentadecan-1-one
SMILES CCCCCCCCCCCCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 50°C
Color White-Yellow
Quantity 5 g
Formula Weight 302.50
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pentadecanophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.