accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOONFFFSFFFOOO(E/Z)

CAS RN 468095-63-8

IUPAC Name: N-[4-(trifluoromethyl)phenyl]-1-({[4-(trifluoromethyl)phenyl]methyl}sulfanyl)-1-{[3,4,5-tris(benzyloxy)-6-[(benzyloxy)methyl]oxan-2-yl]oxy}methanimine
Synonyms: N-[4-(trifluoromethyl)phenyl]-1-({[4-(trifluoromethyl)phenyl]methyl}sulfanyl)-1-{[3,4,5-tris(benzyloxy)-6-[(benzyloxy)methyl]oxan-2-yl]oxy}methanimine
Molecular Weight (g/mol): 901.96
Molecular Formula: C50H45F6NO6S
InChi Key: LNIPCBRVKJJXQX-UHFFFAOYNA-N
SMILES: FC(F)(F)C1=CC=C(CSC(OC2OC(COCC3=CC=CC=C3)C(OCC3=CC=CC=C3)C(OCC3=CC=CC=C3)C2OCC2=CC=CC=C2)=NC2=CC=C(C=C2)C(F)(F)F)C=C1
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)