accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NOHNOHOOHO

CAS RN 4685-12-5

IUPAC Name: 2-(2-aminoacetamido)butanedioic acid
Synonyms: 2-(2-aminoacetamido)butanedioic acid
Molecular Weight (g/mol): 190.16
Molecular Formula: C6H10N2O5
InChi Key: SCCPDJAQCXWPTF-UHFFFAOYNA-N
SMILES: NCC(=O)NC(CC(O)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results