Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycyl-L-aspartic Acid Monohydrate 98.0+%, TCI America™
SDP

Catalog No. G01211G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G01211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G01211G Supplier TCI America Supplier No. G01211G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity dipeptide featuring >98.0% purity for biochemical research applications.

  • Purity (Nonaqueous Titration): ≥98.0% (calculated on anhydrous substance)
  • Appearance: White to almost white powder to crystal
  • Molecular Formula: C6H10N2O5·H2O
  • Molecular Weight: 208.18
  • Specific Rotation: +11.0° to +14.0° ([α]20/D)
  • Water Content: 8.0% to 10.0%
  • Solubility in Water: Almost transparent

Chemical Identifiers

CAS 4685-12-5
Molecular Formula C6H10N2O5
Molecular Weight (g/mol) 190.16
MDL Number MFCD00025585
InChI Key SCCPDJAQCXWPTF-UHFFFAOYNA-N
Synonym H-Gly-Asp-OH
PubChem CID 97363
ChEBI CHEBI:73804
IUPAC Name (2S)-2-[(2-aminoacetyl)amino]butanedioic acid
SMILES C(C(C(=O)O)NC(=O)CN)C(=O)O

Specifications

Color White-Yellow
Packaging Glass Bottle
Quantity 1 g
Formula Weight 190.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Glycyl-L-aspartic Acid Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.