accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4688-76-0

IUPAC Name: {[1,1'-biphenyl]-2-yl}boronic acid
Synonyms: [1,1'-biphenyl]-2-ylboronic acid
Molecular Weight (g/mol): 198.03
Molecular Formula: C12H11BO2
InChi Key: HYCYKHYFIWHGEX-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC=C1C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)