Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biphenyl-2-boronic acid, 95%

Catalog No. AAL1754703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1754703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1754703 Supplier Thermo Scientific Chemicals Supplier No. L1754703
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 4688-76-0 | C12H11BO2 | 198.028 g/mol

It is a substrate employed in a palladium-catalyzed cross-coupling with benzylic acetates providing diarylmethanes. It is a reactant involved in Suzuki-Miyaura cross-coupling reactions with arylhalides, dibromovinyl precursors, alkenyl tosylates and mesylates, and quinoline carboxylates. It is involved in intramolecular Friedel-Crafts alkylation for synthesis of chiral tetralins, hydroxylation to phenols and oxidative coupling with alkynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a substrate employed in a palladium-catalyzed cross-coupling with benzylic acetates providing diarylmethanes. It is a reactant involved in Suzuki-Miyaura cross-coupling reactions with arylhalides, dibromovinyl precursors, alke
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4688-76-0
Molecular Formula C12H11BO2
Molecular Weight (g/mol) 198.028
MDL Number MFCD00136929
InChI Key HYCYKHYFIWHGEX-UHFFFAOYSA-N
Synonym 2-biphenylboronic acid, biphenyl-2-boronic acid, biphenyl-2-ylboronic acid, 2-biphenylboronicacid, 2-phenylphenyl boranediol, 2-phenylphenyl boronic acid, 1,1'-biphenyl-2-ylboronic acid, 2-biphenylylboronic acid, 1,1'-biphenylboronic acid, 2-phenylbenzeneboronic acid
PubChem CID 4589187
IUPAC Name (2-phenylphenyl)boronic acid
SMILES B(C1=CC=CC=C1C2=CC=CC=C2)(O)O

Specifications

Melting Point 173°C to 194°C
Quantity 1 g
Beilstein 3031806
Solubility Information Insoluble in water.
Formula Weight 198.03
Percent Purity 95%
Chemical Name or Material Biphenyl-2-boronic acid
Hazard Category H302-H315-H317-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.