accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 469-83-0

IUPAC Name: (1S,4S,12S,13R,16R,17R)-17-(hydroxymethyl)-12-methyl-8-oxapentacyclo[14.2.1.0¹,¹³.0⁴,¹².0⁵,⁹]nonadeca-5(9),6-dien-17-ol
Synonyms: cafestol
Molecular Weight (g/mol): 316.44
Molecular Formula: C20H28O3
InChi Key: DNJVYWXIDISQRD-HWUKTEKMSA-N
SMILES: C[C@@]12CCC3=C(C=CO3)[C@H]1CC[C@@]13C[C@@H](CC[C@@H]21)[C@@](O)(CO)C3
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (5)
  • (2)
  • (1)
  • (2)
  • (3)