accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4702-13-0

IUPAC Name: 2-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)acetic acid
Synonyms: (1,3-dioxoisoindol-2-yl)acetic acid
Molecular Weight (g/mol): 205.17
Molecular Formula: C10H7NO4
InChi Key: WQINSVOOIJDOLJ-UHFFFAOYSA-N
SMILES: OC(=O)CN1C(=O)C2=CC=CC=C2C1=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)