Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(2-Pyridyldithio)Propionic Acid N-Hydroxysuccinimide Ester

Catalog No. P341525MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mg
100 mg
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P341525MG 100 g
P34155MG 100 mg
P3415100MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P341525MG Supplier Sigma Aldrich Supplier No. P341525MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-(2-Pyridyldithio)Propionic Acid N-Hydroxysuccinimide Ester

Specifications

CAS 4702-13-0
Melting Point 193°C to 196°C
Molecular Formula C10H7NO4
Linear Formula C10H7NO4
MDL Number MFCD00005900
Quantity 100 g
Synonym N-Succinimidyl 3-(2-pyridyldithio)propionate; SPDP
Molecular Weight (g/mol) 205.17 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.