accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3O2H

CAS RN 4712-38-3

IUPAC Name: butan-1-(²H)ol
Synonyms: butan-1-(²H)ol
Molecular Weight (g/mol): 75.13
Molecular Formula: C4H10O
InChi Key: LRHPLDYGYMQRHN-UICOGKGYSA-N
SMILES: [2H]OCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results