Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butanol-d1 97atom%D, TCI America™
SDP

Catalog No. B08255G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B08255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B08255G Supplier TCI America Supplier No. B08255G

Chemical Identifiers

CAS 4712-38-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 75.13
MDL Number MFCD00002965
InChI Key LRHPLDYGYMQRHN-UICOGKGYSA-N
Synonym butanol-d1, butan-1-2h ol, 1-butanol-d, butyl alcohol-d, 1-butanol-od, 1-butanol-d1, 1-butan ol-d, monodeuterobutyl alcohol, butan-1-2 h ol, 1-butyl alcohol-od, 98 atom % d
PubChem CID 20836
IUPAC Name butan-1-(²H)ol
SMILES [2H]OCCCC

Specifications

Melting Point -90°C
Color Colorless
Boiling Point 118°C
UN Number 1120
Quantity 5 g
Formula Weight 75.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Butanol-d1 97atom%D
EINECSNumber (2)-3049
RTECSNumber EO1400000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.