CAS RN 472-93-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
γ-Carotene, ≥95% (HPLC), MilliporeSigma™ Supelco™
γ-Carotene is a plant carotenoid with provitamin A activity.