accessibility menu, dialog, popup

UserName

Viewing profile as user:
HONH

CAS RN 4720-29-0

IUPAC Name: 3-(benzylamino)propan-1-ol
Synonyms: 3-(benzylamino)propan-1-ol
Molecular Weight (g/mol): 165.24
Molecular Formula: C10H15NO
InChi Key: ZQJXSIOFSZYGMH-UHFFFAOYSA-N
SMILES: OCCCNCC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results