accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4721-22-6

IUPAC Name: (6-phosphonohexyl)phosphonic acid
Synonyms: 6-phosphonohexylphosphonic acid
Molecular Weight (g/mol): 246.14
Molecular Formula: C6H16O6P2
InChi Key: WDYVUKGVKRZQNM-UHFFFAOYSA-N
SMILES: OP(O)(=O)CCCCCCP(O)(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results