Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,6-Hexylenediphosphonic Acid 98.0+%, TCI America™
SDP

Catalog No. H15361G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H15361G Supplier TCI America Supplier No. H15361G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4721-22-6
Molecular Formula C6H16O6P2
Molecular Weight (g/mol) 246.136
MDL Number MFCD01630742
InChI Key WDYVUKGVKRZQNM-UHFFFAOYSA-N
Synonym 1,6-Hexanediphosphonic Acid
PubChem CID 2773795
IUPAC Name 6-phosphonohexylphosphonic acid
SMILES C(CCCP(=O)(O)O)CCP(=O)(O)O

Specifications

Melting Point 206°C
Color White
Quantity 1 g
UN Number 3261
Formula Weight 246.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,6-Hexylenediphosphonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.