accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4727-31-5

IUPAC Name: 2-({[1,1'-biphenyl]-4-yl}carbamoyl)benzoic acid
Synonyms: 2-({[1,1'-biphenyl]-4-yl}carbamoyl)benzoic acid
Molecular Weight (g/mol): 317.34
Molecular Formula: C20H15NO3
InChi Key: SLUINPGXGFUMLL-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1C(=O)NC1=CC=C(C=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results