accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CPOOHHO

CAS RN 4730-79-4

IUPAC Name: nonylphosphonic acid
Synonyms: nonylphosphonic acid
Molecular Weight (g/mol): 208.24
Molecular Formula: C9H21O3P
InChi Key: OLGGYSFJQGDOFX-UHFFFAOYSA-N
SMILES: CCCCCCCCCP(O)(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results