Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonylphosphonic Acid 98.0+%, TCI America™
SDP

Catalog No. N10251G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N10251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N10251G Supplier TCI America Supplier No. N10251G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4730-79-4
Molecular Formula C9H21O3P
Molecular Weight (g/mol) 208.238
MDL Number MFCD00015842
InChI Key OLGGYSFJQGDOFX-UHFFFAOYSA-N
PubChem CID 17843951
IUPAC Name nonylphosphonic acid
SMILES CCCCCCCCCP(=O)(O)O

Specifications

Melting Point 104°C
Color White-Yellow
Quantity 1 g
UN Number 3261
Formula Weight 208.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nonylphosphonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.