accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOHOH2N

CAS RN 4733-65-7

IUPAC Name: 3-carbamoylpyridine-2-carboxylic acid
Synonyms: 3-carbamoylpyridine-2-carboxylic acid
Molecular Weight (g/mol): 166.14
Molecular Formula: C7H6N2O3
InChi Key: CGDAWEVRGAIHJH-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(N=CC=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results