accessibility menu, dialog, popup

UserName

Viewing profile as user:
NN

CAS RN 4734-49-0

IUPAC Name: 5-(piperidin-1-yl)pentanenitrile
Synonyms: 5-(piperidin-1-yl)pentanenitrile
Molecular Weight (g/mol): 166.27
Molecular Formula: C10H18N2
InChi Key: FDSRLFIEZWQEEU-UHFFFAOYSA-N
SMILES: N#CCCCCN1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results