Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Piperidinovaleronitrile, Thermo Scientific™

Catalog No. AC147650250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC147650250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC147650250 Supplier Thermo Scientific Chemicals Supplier No. 147650250

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4734-49-0
Molecular Formula C10H18N2
Molecular Weight (g/mol) 166.268
InChI Key FDSRLFIEZWQEEU-UHFFFAOYSA-N
Synonym 5-piperidinovaleronitrile, piperidine-1-valeronitrile, 5-piperidin-1-ylvaleronitrile, 5-piperidin-1-yl pentanenitrile, piperidinepentanenitrile, 1-piperidinevaleronitrile, 4-piperidinobutyl cyanide, 1-piperidinepentanenitrile, 5-1-piperidinyl pentanenitrile
PubChem CID 78472
IUPAC Name 5-piperidin-1-ylpentanenitrile
SMILES C1CCN(CC1)CCCCC#N

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Brown
Density 0.91g/mL
Infrared Spectrum Authentic
Assay Percent Range 96%
Refractive Index 1.4660 to 1.4680 (20°C, 589nm)
Quantity 25 g
Beilstein 20, IV, 1067
Formula Weight 166.27
Percent Purity ≥95%
Physical Form Liquid
Chemical Name or Material 5-Piperidinovaleronitrile, 96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.