accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4736-60-1

IUPAC Name: ethyltriphenylphosphanium iodide
Synonyms: ethyl(triphenyl)phosphonium iodide
Molecular Weight (g/mol): 418.26
Molecular Formula: C20H20IP
InChi Key: SLAFUPJSGFVWPP-UHFFFAOYSA-M
SMILES: [I-].CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (8)
Quantity 
  • (3)
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (6)
  • (1)
  • (8)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)