Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyltriphenylphosphonium iodide, Thermo Scientific™

Catalog No. AA3041236
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3041236 500 g
AA3041214 25 g
AA3041222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3041236 Supplier Thermo Scientific Chemicals Supplier No. 3041236

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4736-60-1
Molecular Formula C20H20IP
Molecular Weight (g/mol) 418.26
MDL Number MFCD00040352
InChI Key SLAFUPJSGFVWPP-UHFFFAOYSA-M
Synonym ethyltriphenylphosphonium iodide, phosphonium, ethyltriphenyl-, iodide, ethyl triphenylphosphonium iodide, phenylphosphonium ethyl iodide, unii-xxz2v239jn, xxz2v239jn, ethyltriphenylphosphanium iodide, phosphonium, ethyltriphenyl-, iodide 1:1, ethyl triphenyl phosphanium iodide, ethyl triphenyl phosphonium iodide
PubChem CID 78474
IUPAC Name ethyltriphenylphosphanium iodide
SMILES [I-].CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 164°C to 165°C
Odor Odorless
Quantity 500 g
UN Number UN2811
Formula Weight 418.26
Physical Form Powder
Chemical Name or Material Ethyltriphenylphosphonium iodide
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P301+P310-P305+P351+P338-P362-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.