accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNClCl

CAS RN 47369-00-6

IUPAC Name: 1,1'-diphenyl-1λ⁵,1'λ⁵-[4,4'-bipyridine]-1,1'-bis(ylium) dichloride
Synonyms: 1,1'-diphenyl-1λ⁵,1'λ⁵-[4,4'-bipyridine]-1,1'-bis(ylium) dichloride
Molecular Weight (g/mol): 381.3
Molecular Formula: C22H18Cl2N2
InChi Key: SZZVUWLOZBMPLX-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].C1=CC=C(C=C1)[N+]1=CC=C(C=C1)C1=CC=[N+](C=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results