Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Diphenyl-4,4'-bipyridinium Dichloride 97.0+%, TCI America™
SDP

Catalog No. D21651G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D21651G Supplier TCI America Supplier No. D21651G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laboratory reagent with 97.0% minimum purity.

Chemical Identifiers

CAS 47369-00-6
Molecular Formula C22H18Cl2N2
Molecular Weight (g/mol) 381.30
MDL Number MFCD00142355
InChI Key SZZVUWLOZBMPLX-UHFFFAOYSA-L
PubChem CID 10861840
IUPAC Name 1,1'-diphenyl-1λ⁵,1'λ⁵-[4,4'-bipyridine]-1,1'-bis(ylium) dichloride
SMILES [Cl-].[Cl-].C1=CC=C(C=C1)[N+]1=CC=C(C=C1)C1=CC=[N+](C=C1)C1=CC=CC=C1

Specifications

Quantity 1 g
Formula Weight 381.30
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Diphenyl-4,4′-bipyridinium Dichloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.