accessibility menu, dialog, popup

UserName

Viewing profile as user:
HN(S)OOOOHOCH3CH3CH3H3CCH3CH3

CAS RN 47375-34-8

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-[4-(tert-butoxy)phenyl]propanoic acid
Synonyms: (2S)-3-[4-(tert-butoxy)phenyl]-2-[(tert-butoxycarbonyl)amino]propanoic acid
Molecular Weight (g/mol): 337.42
Molecular Formula: C18H27NO5
InChi Key: ZEQLLMOXFVKKCN-YQTOOIBONA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(OC(C)(C)C)C=C1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)