Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-O-tert-butyl-L-tyrosine 98.0+%, TCI America™
SDP

Catalog No. B34855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34855G 5 g
B34851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34855G Supplier TCI America Supplier No. B34855G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 47375-34-8
Molecular Formula C18H27NO5
Molecular Weight (g/mol) 337.42
MDL Number MFCD00065598
InChI Key ZEQLLMOXFVKKCN-YQTOOIBONA-N
Synonym boc-tyr tbu-oh, boc-o-tert-butyl-l-tyrosine, boc-tyr tbul-oh, s-3-4-tert-butoxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, n-tert-butoxycarbonyl-o-tert-butyl-l-tyrosine, l-tyrosine, n-1,1-dimethylethoxy carbonyl-o-1,1-dimethylethyl, 2s-3-4-tert-butoxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, 2s-2-tert-butoxy carbonylamino-3-4-tert-butoxy phenyl propanoic acid, boc-tyr but-oh, pubchem12288
PubChem CID 7017901
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-[4-(tert-butoxy)phenyl]propanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(OC(C)(C)C)C=C1)C(O)=O

Specifications

Melting Point 117°C
Color White
Quantity 5 g
Formula Weight 337.42
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-O-tert-butyl-L-tyrosine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.