accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4743-58-2

IUPAC Name: 2-(3-oxo-1,3-dihydro-2-benzofuran-1-yl)acetic acid
Synonyms: (3-oxo-1H-2-benzofuran-1-yl)acetic acid
Molecular Weight (g/mol): 192.17
Molecular Formula: C10H8O4
InChi Key: FJWKEFBYCZSVNZ-UHFFFAOYNA-N
SMILES: OC(=O)CC1OC(=O)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results