accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOBr

CAS RN 4753-59-7

IUPAC Name: 4-bromobutyl acetate
Synonyms: 4-bromobutyl acetate
Molecular Weight (g/mol): 195.06
Molecular Formula: C6H11BrO2
InChi Key: UOABIRUEGSGTSA-UHFFFAOYSA-N
SMILES: CC(=O)OCCCCBr
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)