Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Bromobutyl acetate

Catalog No. 22757950G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-757-950G 50 g
22-757-910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-757-950G Supplier Sigma Aldrich Supplier No. 22757950G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Bromobutyl acetate was used in the synthesis of 9-alkylguanine derivative.

Specifications

CAS 4753-59-7
Density 1.348 g/mL (at 25°C (literature))
Boiling Point 92°C to 93°C (12 mmHg)
Molecular Formula C6H11BrO2
Linear Formula CH3CO2(CH2)4 Br
Refractive Index n20/D 1.46 (literature)
MDL Number MFCD00000263
Quantity 50 g
Molecular Weight (g/mol) 195.05
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.