accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4755-50-4

IUPAC Name: 4-(ethylamino)benzoyl chloride
Synonyms: 4-(ethylamino)benzoyl chloride
Molecular Weight (g/mol): 183.64
Molecular Formula: C9H10ClNO
InChi Key: XFLQAKBFRUFBJS-UHFFFAOYSA-N
SMILES: CCNC1=CC=C(C=C1)C(Cl)=O
Percent Purity 
  • (4)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)