Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ 4-(Dimethylamino)benzoyl chloride, 99.0%, MilliporeSigma™ Supelco™

Catalog No. 111018361 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8361 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8361 Supplier MilliporeSigma Supelco Supplier No. 679541G
Add to Cart
Add to Cart

Material is reported to be very moisture sensitive. Handle under an inert atmosphere

TRUSTED_SUSTAINABILITY

Specifications

CAS 4755-50-4
Product Line LiChropur
Linear Formula (CH3)2 NC6H4 COCl
MDL Number MFCD00236039
Melting Point 145°C to 149°C; 146°C to 150°C
Synonym DMABC
Type Organic Standard
Quantity 1 g
Formula Weight 183.63
Percent Purity 99.0 to 101% (AT)
Grade HPLC Derivatization
Show More Show Less

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.