accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 476-60-8

IUPAC Name: anthracene-1,4,9,10-tetrol
Synonyms: anthracene-1,4,9,10-tetrol
Molecular Weight (g/mol): 242.23
Molecular Formula: C14H10O4
InChi Key: BKNBVEKCHVXGPH-UHFFFAOYSA-N
SMILES: OC1=C2C(O)=C3C=CC=CC3=C(O)C2=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results